Anti-PCSK2 Mouse Monoclonal Antibody [clone: 3H4]
Catalog # ABNOH00005126-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Proprotein convertase subtilisin/kexin type 2
- Antigen symbol:PCSK2
- Clonality:Monoclonal
- Clone:3H4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5126
- Antigen synonyms:Proprotein convertase 2|PC2|NEC 2|NEC2|Neuroendocrine convertase 2|KEX2-like endoprotease 2|PCSK2|Prohormone convertase 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PCSK2
Clonality: Monoclonal
Clone: 3H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human