Anti-PAEP Mouse Monoclonal Antibody [clone: 4E4]
ABNOH00005047-M02
: Abnova
- Antibody type:Primary
- Antigen name:progestagen-associated endometrial protein
- Antigen symbol:PAEP
- Clonality:Monoclonal
- Clone:4E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5047
- Antigen synonyms:PEP|GD|GdA|GdS|GdF|PAEG|PP14
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PAEP
Clonality: Monoclonal
Clone: 4E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human