Anti-MEIS2 Mouse Monoclonal Antibody [clone: 1D1]
ABNOH00004212-M03
: Abnova
- Antibody type:Primary
- Antigen name:Meis homeobox 2
- Antigen symbol:MEIS2
- Clonality:Monoclonal
- Clone:1D1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4212
- Antigen synonyms:HsT18361|Meis homeobox 2|TALE homeobox protein Meis2|Meis1|OTTHUMP00000160209|MRG1|myeloid ecotropic viral integration site 1 homolog 2|Meis homolog 2|Meis1-related gene 1|MGC2820|MEIS2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: MEIS2
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human