Anti-LDHC Mouse Monoclonal Antibody [clone: 2C8]
ABNOH00003948-M01
: Abnova
- Antibody type:Primary
- Antigen name:Lactate dehydrogenase C
- Antigen symbol:LDHC
- Clonality:Monoclonal
- Clone:2C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3948
- Antigen synonyms:CT32|LDHX|LDH3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: LDHC
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human