Anti-GRM8 Mouse Monoclonal Antibody [clone: 4A7]
Catalog # ABNOH00002918-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor, metabotropic 8
- Antigen symbol:GRM8
- Clonality:Monoclonal
- Clone:4A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2918
- Antigen synonyms:GLUR8|mGlu8|GPRC1H|MGLUR8
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GRM8
Clonality: Monoclonal
Clone: 4A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human