Order Entry
Poland
ContactUsLinkComponent
Anti-ESD Mouse Monoclonal Antibody [clone: 1E1]
Anti-ESD Mouse Monoclonal Antibody [clone: 1E1]
  ABNOH00002098-M01
 :  Abnova
undefined
Anti-ESD Mouse Monoclonal Antibody [clone: 1E1]
  ABNOH00002098-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Esterase D
  • Antigen symbol:
    ESD
  • Clonality:
    Monoclonal
  • Clone:
    1E1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    2098
  • Antigen synonyms:
    FGH
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: ESD
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human