Anti-RPL37 Rabbit Polyclonal Antibody
Catalog # ABGEAP9565B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal Protein L37
- Antigen symbol:RPL37
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Gene ID:55361
- Antigen synonyms:PIK42A|Phosphatidylinositol 4-kinase type II|PI4KII|Type II alpha phosphatidylinositol 4-kinase
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:YELFKKDPGFDRGQC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This PI4K2A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 387-415 amino acids from the C-terminal region of human PI4K2A.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: RPL37 (ribosomal protein L37)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human