Anti-CDK4 Rabbit Polyclonal Antibody
ABGEAP7520B
: Abgent
- Antibody type:Primary
- Antigen name:Cyclin-dependent Kinase 4
- Antigen symbol:CDK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal (250–350 aa)
- Cross adsorption:No
- Antigen synonyms:CMM3|MGC14458|PSK-J3|Melanoma Cutaneous Malignant 3|Cell Division Kinase 4|Cell Division Protein Kinase 4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CDK4 (cyclin-dependent kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human