Anti-CAPN9 Rabbit Polyclonal Antibody
Catalog # ABGEAP7310A
Supplier: Abgent
Jump to:
Product Details & Documents
Type: Primary
Antigen: CAPN9 (calpain 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Documents
AP7310a.pdf
Datasheet
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Epitope:N-terminal
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:Calpain 9
- Cat. No.:ABGEAP7310A
- Reactivity:Human
- Antigen synonyms:nCL-4|Calpain-9|New calpain 4|NCL4|CAPN9|Digestive tract-specific calpain|Protein CG36
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:CAPN9
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



