Order Entry
Poland
ContactUsLinkComponent
 
Anti-UBE3A Rabbit Polyclonal Antibody
  ABGEAP2154B
 :  Abgent
undefined
Anti-UBE3A Rabbit Polyclonal Antibody
  ABGEAP2154B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin Protein Ligase E3A
  • Antigen symbol:
    UBE3A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Antigen synonyms:
    Human papillomavirus E6-associated protein|Ubiquitin-protein ligase E3A|E6-AP|ANCR|EPVE6AP|AS|Oncogenic protein-associated protein E6-AP|E6AP ubiquitin-protein ligase|HPVE6A
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: UBE3A (ubiquitin protein ligase E3A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human