Anti-CCT4 Rabbit Polyclonal Antibody
ABGEAP14904A
: Abgent
- Antibody type:Primary
- Antigen name:Chaperonin Containing TCP1, subunit 4 (delta)
- Antigen symbol:CCT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal
- Gene ID:10575
- Antigen synonyms:SRB|Cctd|CCT-DELTA
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:57.924 kDa
- Sequence:RGKGAYQDRDKPAQC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CCT4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 4-32 amino acids from the N-terminal region of human CCT4.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: CCT4 (chaperonin containing TCP1, subunit 4 (delta))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human