Anti-HMFG Rabbit Polyclonal Antibody
ABGEAP12888B
: Abgent
- Antibody type:Primary
- Antigen name:Milk Fat Globule-EGF Factor 8 Protein
- Antigen symbol:MFGE8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:SPAG10|EDIL1|MFG-E8|BA46|OAcGD3S|hP47|MFGM|HsT19888|SED1|HMFG
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: HMFG
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human