Anti-PMM2 Rabbit Polyclonal Antibody
Catalog # AVIVARP63939_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:phosphomannomutase 2
- Antigen symbol:PMM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5373
- Antigen synonyms:PMM 2|PMI|CDGS|CDG1|PMI1|CDG1a
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PMM2 (phosphomannomutase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: