Anti-ADAM33 Rabbit Polyclonal Antibody
AVIVARP49937_P050
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase domain 33
- Antigen symbol:ADAM33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:80332
- Antigen synonyms:DJ964F7.1|C20orf153
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ADAM33 (ADAM metallopeptidase domain 33)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: