Anti-CHERP Rabbit Polyclonal Antibody
AVIVARP40642_P050
- Antibody type:Primary
- Antigen name:Calcium Homeostasis Endoplasmic Reticulum Protein
- Antigen symbol:CHERP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:10523
- Antigen synonyms:DAN16|SRA1|SCAF6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CHERP (calcium homeostasis endoplasmic reticulum protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: