Anti-ANXA3 Rabbit Polyclonal Antibody
Catalog # AVIVARP36578_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:annexin A3
- Antigen symbol:ANXA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:306
- Antigen synonyms:ANX3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ANXA3 (annexin A3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: