Anti-BCAM Rabbit Polyclonal Antibody
Catalog # BOSSBS-7546R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Basal Cell Adhesion Molecule
- Antigen symbol:BCAM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:MSK19|F8/G253 antigen|B-cell adhesion molecule|CD239|AU|B-CAM cell surface glycoprotein|lutheran antigen|Auberger b antigen|LU|lutheran blood group glycoprotein
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: BCAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat