Tubing clamps, spring style, SP Bel-Art
Stainless steel/acetal plastic. This positive-action, self-closing clamp opens by squeezing the handles together.
- Perfect for all flexible tubing
- Chemical and stain-resistant
- Fits tubing up to Ø 8 or 12 mm
Frequently Bought Together