Order Entry
Poland
ContactUsLinkComponent
 
Titrators, automatic, TitraLab® AT1000
 :  Hach
 
undefined
Titrators, automatic, TitraLab® AT1000
 :  Hach

 

  • Catalog No:
  • 661-0199
  • 661-0202
  • 661-0200
  • 661-0201
  • Model:
  • TitraLab® AT1000
  • TitraLab® AT1000
  • TitraLab® AT1000
  • TitraLab® AT1000
  • Display:
  • Graphic colour, VGA, 5,7"
  • Graphic colour, VGA, 5,7"
  • Graphic colour, VGA, 5,7"
  • Graphic colour, VGA, 5,7"
  • Keypad:
  • Soft keypad
  • Soft keypad
  • Soft keypad
  • Soft keypad
  • Dosing volume:
  • 2,5/5/10/25 ml
  • 2,5 / 5 / 10 / 25 ml
  • 2,5 / 5 / 10 / 25 ml
  • 2,5 / 5 / 10 / 25 ml
  • Dosing speed:
  • 170 ml/min
  • 170 ml/min
  • 170 ml/min
  • 170 ml/min
  • Burette resolution:
  • 20000 steps
  • 20000 steps
  • 20000 steps
  • 20000 steps
  • pH resolution:
  • ±0,001
  • ±0,001
  • ±0,001
  • ±0,001
  • mV resolution:
  • ±0,1
  • ±0,1
  • ±0,1
  • ±0,1
  • Measuring input electrode:
  • INTELLICAL (“Plug & Play” digital), analogue or photocolorimetric electrodes
  • INTELLICAL (“Plug & Play” digital), analogue or photocolorimetric electrodes
  • INTELLICAL (“Plug & Play” digital), analogue or photocolorimetric electrodes
  • INTELLICAL (“Plug & Play” digital), analogue or photocolorimetric electrodes
  • Connections:
  • 2 × electrode ports, 2 × USB ports, 1 × serial port, 1 × ethernet
  • 2 × electrode ports, 2 × USB ports, 1 × serial port, 1 × ethernet
  • 2 × electrode ports, 2 × USB ports, 1 × serial port, 1 × ethernet
  • 2 × electrode ports, 2 × USB ports, 1 × serial port, 1 × ethernet
  • Power supply:
  • 240 V, 50/60 Hz
  • 240 V, 50/60 Hz
  • 240 V, 50/60 Hz
  • 240 V, 50/60 Hz
  • W×D×H:
  • 400×360×220 mm
  • 400×360×220 mm
  • 400×360×220 mm
  • 400×360×220 mm
  • Weight:
  • 4 kg
  • 4 kg
  • 4 kg
  • 4 kg

 

Water application packs

 

Frequently Bought Together