Donor horse serum
Supplier: Biowest
Certificates
About this item
Donor horse serum is aseptically collected from herds of healthy animals inspected periodically by a licensed veterinarian. Applications for donor horse serum include mycoplasma contamination tests carried out on media, blocking agent and controls in immunoassays, culture of PC12 cells when combined with FBS.
Frequently Bought Together