Order Entry
Poland
ContactUsLinkComponent
 
3-Bromohexane ≥80.0% (by GC) (contains 2-Bromohexane) stabilised
  TCIAB1289-10G
 :  TCI
 :  
undefined
3-Bromohexane ≥80.0% (by GC) (contains 2-Bromohexane) stabilised
  TCIAB1289-10G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236
  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 80.0 %

 

 

 :
 :165.07 g/mol
 :222-174-9
 :1993
 :GHS02
 :3377-87-5
 :H₃CCH₂BrCH(CH₂)₂CH₃
 :MFCD00039181
 :142°C
 :Stabilised with copper chip
 :46°C
 :3 III