Hexane (mixture of isomers)
CHMP424664000.9060
: CHEMPUR
:
- Pack type:Plastic bottle
- Pk:60 L
- Seq:5407
- Environmentally Preferable:
: Hexanes, Isohexane
:
:86.18 g/mol
:203-777-6
:1208
:GHS02
:GHS08
:GHS07
:GHS09
:–22 °C
:Ambient
:110-54-3
:92112-69-1
:H₃C(CH₂)₄CH₃
:MFCD02179311
:68…70 °C (1013 hPa)
:–95 °C
:0.66 g/cm³ (20 °C)
:3 II
:13,04712