(±)-Taxifolin ≥90% (by HPLC), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Taxifolin, Dihydroquercetin, 3,3′,4′,5,7-Pentahydroxyflavanone
CAS Number:24198-97-8
:480-18-2
Formula:C₁₅H₁₂O₇
MDL Number:MFCD11044454
Molecular Weight:304.25 g/mol (anhy)
:M-Clarity™ quality level = MQ100
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together