Gellan gum
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:71010-52-1
:
MDL Number:MFCD00131909
Storage Temperature:Ambient
EINECS:275-117-5
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberALFAJ63423.30_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Thermo Scientific Cell Culture Workflow
Product Literature
Frequently Bought Together