Sample tubes for Roche Cobas analysers
SIMPB721-1
- Packed:Bag of 1000, Carton of 2000
- Capacity:2,5 ml
- Description:For Roche Cobas analysers, without caps
- Environmentally Preferable:
- Pk:2.000
PS, clear.
- Good chemical resistance
- Good temperature resistance
This tube is used with the Roche Cobas Analyzer. It has perfect clarity for easy viewing of contents. This graduated false bottom tube has a capacity of 2,5 ml and will accept bar code labels.