Antibiotic assay discs
Supplier: MACHEREY-NAGEL
Certificates
About this item
These products are used in testing the resistance of pathogens to antibiotics. For this test the filter paper sections can be impregnated with the antibiotic to be tested and placed on the inoculated nutrient medium. Depending on the effectiveness, a smaller or larger zone of inhibition is formed.
- Non-impregnated filter paper sections
- Basis weight: 270 g/m²
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together