Menadione sodium bisulphite reference standard, Dr. Ehrenstorfer
Catalog # EHERC14863200
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:0,25 g
- CAS No.:130-37-0
- Description:Menadione sodium bisulphite reference standard
- Environmentally Preferable:
- Shipping conditions:Ambient
- Synonym(s):2-Naphthalenesulfonic acid, 1,2,3,4-tetrahydro-2-methyl-1,4-dioxo-, sodium salt (8CI,9CI), 1,2,3,4-Tetrahydro-2-methyl-1,4-dioxo-2-naphthalenesulfonic acid sodium salt, 1,4-Naphthalenedione, 2-methyl-, sodium bisulfite deriv., 2,3-Dihydro-2-methyl-1,4-naphthoquinone-2-sulfonate sodium, 2-Methyl-1,4-naphthoquinone sodium bisulfite, 2-Methyl-1,4-naphthoquinone sodium hydrogen sulfite, 2-Methylnaphthoquinone sodium hydrogen sulfite, Golagen K, Hemoklot, Hetrogen K, Hetrogen K Premix, Hykinone, Ido-K, K-Trombina, Kalzon, Kavitamin, Kavitan, Kawitan, Klotogen, Klotogen F, Klotogen F 16, Klotogen F 227, MSBC, Menadione bisulfite, Menadione sodium bisulfite, Menadione sodium hydrogen sulfite, Menaphthone sodium bisulfite, Menaphthone sodium bisulphite, Sodium menadione bisulfite, Vikasol, Vitamin K injection, Vitamin K3 sodium bisulfite, 2-Naphthalenesulfonic acid, 1,2,3,4-tetrahydro-2-methyl-1,4-dioxo-, sodium salt (1:1)
- Formula:C11H9O5S.Na
- Molecular weight:276.24 g/mol
- Storage temperature:-18°C
- Appearance:Neat
- Form:Neat