Test tubes with or without rim, heavy wall
Supplier: GLASWARENFABRIK KARL HECHT
Certificates
About this item
Thick-walled, made of borosilicate glass 3.3, exceptional mechanical strength.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Frequently Bought Together