Auramine O, pure biological stain
Catalog # ACRO227520500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to ochre Powder
- Infrared spectrum:Conforms
- Dye content:≥60 %
Specification Test Results
About this item
Synonyms: Pyoctanunum aureum, 4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Pyoktanin Yellow, 4,4'-(Iminomethylene)bis(N,N-dimethylaniline) hydrochloride, Basic Yellow 2, Canary Yellow
:
:303.84 g/mol
EINECS:219-567-2
UN:2811
:GHS06
:GHS08
Storage Temperature:Ambient
CAS Number:2465-27-2
Formula:C₁₇H₂₂ClN₃
MDL Number:MFCD00012484
Melting Pt:>250 °C (dec.)
ADR:6.1 III