Order Entry
Netherlands
ContactUsLinkComponent
Potassium bromate/Potassium bromide 0,0167 M in aqueous solution
Potassium bromate/Potassium bromide 0,0167 M in aqueous solution
Catalog # REGEKB20016W
Supplier:  REAGECON
CAS Number:  
Potassium bromate/Potassium bromide 0,0167 M in aqueous solution
Catalog # REGEKB20016W
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

About this item

 :
EINECS:231-829-8
 :GHS08
Storage Temperature:Ambient
CAS Number:7758-01-2
Formula:KBrO₃/KBr
MDL Number:MFCD00011359