1,2,3,6-Tetrahydrophthalic anhydride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 4-Cyclohexene-1,2-dicarboxylic anhydride
CAS Number:85-43-8
:
Formula:C₈H₈O₃
Molecular Weight:152.15 g/mol
ADR:8 III
:M-Clarity™ quality level = MQ200
EINECS:201-605-4
UN:2698
:GHS08
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:10 g/l (20 °C)
- Melting Point:103 °C
- Molar Mass:152.14 g/mol
- Boiling Point:195 °C (67 hPa)
- Vapor Pressure:0.013 hPa (20 °C)
- Flash Point:156 °C
- Density:1.375 g/cm3 (20 °C)
- pH:2.1 (10 g/l, H2O, 20 °C)
- Ignition Point:400 °C
Specification Test Results
Frequently Bought Together