1,2,4,5-Tetrabromobenzene 97%
:636-28-2
:
:C₆H₂Br₄
:MFCD00000063
:393.7 g/mol
:177…182 °C
:211-253-3
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:White to beige to brown Powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:177 to 183 °C
- GC:≥96.0 %
Frequently Bought Together