β-Naphthoflavone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 5,6-Benzoflavone, 3-Phenyl-1H-naphtho[2,1-b]pyran-1-one, 3-Phenyl-1H-benzo[f]chromen-1-one
CAS Number:6051-87-2
:
Formula:C₁₉H₁₂O₂
MDL Number:MFCD00004986
Boiling Pt:461
Melting Pt:162…166 °C
Density:1.11 g/cm³ (20 °C)
Molecular Weight:272.3 g/mol
Storage Temperature:Ambient
EINECS:227-958-4
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together