Order Entry
Netherlands
ContactUsLinkComponent
 
Bromine ≥99%, extra pure, Supelco®
  1.01945.1000
 :  Merck
 :  
Bromine ≥99%, extra pure, Supelco®
  1.01945.1000
 :  Merck
 :  

 

  • Pack type:
    Glass bottle, plastic-coated, returnable
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

 

 

  • Assay (iodometric):
    ≥ 99 %
  • Identity:
    passes test
  • Iodine (I):
    ≤ 0.002 %
  • Organic bromine compounds:
    passes test
  • Non volatile matter:
    ≤ 0.02 %

 

 

 :
 :159.81 g/mol
 :231-778-1
 :1744
 :GHS06
 :GHS05
 :GHS09
 :Ambient
 :7726-95-6
 :Br₂
 :MFCD00010896
 :58.7
 :–7.2 °C
 :3.12 g/cm³ (20 °C)
 :8 (6.1) I
 :13,01378
 :M-Clarity™ quality level = MQ100