Beryllium ≥99.7% (metals basis), wire, annealed, Ø 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-41-7
:
Formula:Be
MDL Number:MFCD00134032
Boiling Pt:2970
Melting Pt:1278
Density:1.85 g/cm³ (20 °C)
Molecular Weight:9.01 g/mol
Merck Index:12,01201
EINECS:231-150-7
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together