Bottle-top dispensers, seripettor® / seripettor® pro
Supplier: Brand
Certificates
About this item
Variable volume.
- Easily replaceable cartridge and seal
- Automatic dispensing: Spring-loaded piston refills the cylinder after use
- Volume setting: Indented track for quick and exact volume setting
- Can be taken apart without tools, easy cleaning and maintenance
- seripettor® for alkaline, buffer solutions and culture media, e.g. agar nutrient medium, up to +60 °C
- seripettor® pro for dispensing many acidic, alkaline and saline solutions, as well as solvents such as essential oils
The seripettor® and seripettor® pro with innovative and economic design are suitable for daily use. Precise, robust and simple to use, they are used in many applications. Easy maintenance and cleaning, the entire dispensing unit can be quickly exchanged by the user, without tools (no expensive repairs).
Ordering information: Supplied with a discharge tube, a filling tube, one spare dispensing unit and PP adapters (GL 45/32 and GL 45/S40).
Related Information
Brand seripettor® and seripettor® pro Brand Volumetric Measurement in the Laboratory
Product Literature
GA seripettor.pdf
Manual
Liquid Handling with Bottle-Top Instruments
Application Notes
Frequently Bought Together