Lens cleaning tissues, Grade 105, Whatman™
These cleaning tissues are suitable for use with lenses and any other optical surfaces made from glass, quartz or plastic.
- Soft surface
- Strong and highly absorbent
- Chemically pure tissue free from silicones and other additives
Frequently Bought Together