Safety spectacles, Millennia™ Classic
Comfortable safety spectacles with excellent optical qualities for complete protection.
- Straight temples, no adjustments necessary
- Comfortable, universal nose bridge
- Easily replaceable lenses
- Adjustable, break-away Flexicord retainer for added security and convenience
Weight: 32 g
EN 166, EN 170, EN 171, EN 172 (*); 1F
: EN 166, EN 170, EN 171, EN 172 (*); 1F
Frequently Bought Together