Guaifenesin
Catalog # PROOMM0354.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
Synonyms: Guaiacol glyceryl ether, 3-(2-Methoxyphenoxy)-1,2-propanediol, 3-(2-Methoxyphenoxy)propane-1,2-diol
:
:198.22 g/mol
EINECS:202-222-5
CAS Number:93-14-1
Formula:C₁₀H₁₄O₄
MDL Number:MFCD00016873