About this item
Sox genes comprise a family of genes that are related to the mammalian sex-determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.
Type: Primary
Antigen: SOX4
Clonality: Polyclonal
Clone:
Conjugation: AbBy Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken
Specifications
- Catalog No:
- BOSSBS-11208R-BF55
- BOSSBS11208RBF555T
- Antigen name:
- SOX4
- SOX4
- Antigen synonyms:
- SRY related HMG box gene 4|SRY box containing gene 4|SOX-4|Ecotropic viral integration site 16|EVI16|SOX 4|EVI 16|SRY sex determining region Y box 4
- SRY related HMG box gene 4|SRY box containing gene 4|SOX-4|Ecotropic viral integration site 16|EVI16|SOX 4|EVI 16|SRY sex determining region Y box 4
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- AbBy Fluor® 555
- AbBy Fluor® 555
- Reactivity:
- Chicken, Human, Rat, Mouse
- Chicken, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 6659
- 6659
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Purification:
- Purified by protein A
- Purified by protein A
- Concentration:
- 1 µg/µl
- 1 µg/µl
- Storage buffer:
- Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:
- Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.