HyClone™ Standard Fetal Bovine Serum (FBS), U.S. Origin, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
Certificates
About this item
HyClone™ standard cell culture grade FBS is filtered through three sequential 100 µm (0,1 µm) pore-size rated filters.
- Sourced in the USA with complete traceability back to original source
- Low in antibodies and high in growth factors
- Produced using true pool technology for uniformity and consistency from bottle to bottle
- Filtered through three sequential 100 nm (0.1 µm) pore size-rated filters
HyClone Standard Fetal Bovine Serum (FBS), U.S. origin is an economical alternative to characterized and defined FBS. Standard FBS is ideal for research applications.
Standard FBS is assayed for gamma globulin, alkaline phosphatase, lactate dehydrogenase, glutamic pyruvic transaminase, glutamic oxaloacetic transaminase, pH, total protein, albumin, blood urea nitrogen, creatinine, total bilirubin, sodium, potassium, calcium, chloride, inorganic phosphorus, osmolality, iron, total iron binding capacity (TIBC) and IgG. Included assays are subject to change without notice.
Certifications: Virus panel testing according to 9 CFR 113.53.
Specifications
- Catalog No:
- HYCLSH30088.03HI
- SH30088.03IR25
- HYCLSH30088.02
- HYCLSH30088.02HI
- HYCLSH30088.03
- Origin:
- US
- US
- US
- US
- US
- Storage temperature:
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
Specifications
Frequently Bought Together