Fluoxetine hydrochloride ≥99% (by TLC)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Synonyms: N-Methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propylamine Hydrochloride
CAS Number:56296-78-7
:59333-67-4
Formula:C₁₇H₁₈F₃NO·HCl
MDL Number:MFCD00214288
Molecular Weight:345.79 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together