Benzopinacol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,1,2,2-Tetraphenyl-1,2-ethanediol, 1,1,2,2-Tetraphenylethane-1,2-diol, Tetraphenylethylene glycol, Tetraphenyl-1,2-ethanediol
CAS Number:464-72-2
:
Formula:(C₆H₅)₂C(OH)C(OH)(C₆H₅)₂
MDL Number:MFCD00004448
Boiling Pt:508
Melting Pt:174…180 °C
Density:1.24 g/cm³ (20 °C)
Molecular Weight:366.46 g/mol
Merck Index:13,01101
Storage Temperature:Ambient
EINECS:207-356-8
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together