Gallium metal ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-55-3
:
Formula:Ga
MDL Number:MFCD00134045
Boiling Pt:2403
Melting Pt:29
Density:5.904 g/cm³ (20 °C)
Molecular Weight:69.72 g/mol
EINECS:231-163-8
ADR:8 III
Merck Index:12,04364
UN:2803
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together