3-Bromofuran 98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22037-28-1
:
Formula:C₄H₃BrO
MDL Number:MFCD00005347
Boiling Pt:103 °C (750 mmHg)
Density:1.635 g/cm³
:Stabilised with calcium carbonate
Molecular Weight:146.97 g/mol
Flash Pt:3 °C
Storage Temperature:Freezer
EINECS:244-744-6
UN:1993
ADR:3 II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4955 to 1.4985 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:0.5 % calcium carbonate
Specification Test Results
Frequently Bought Together