S(-)-Proline, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
CAS Number:147-85-3
:
Formula:C₅H₉NO₂
MDL Number:MFCD00064318
Boiling Pt:245
Melting Pt:220…222 °C
Density:1.35 g/cm³ (25 °C)
Molecular Weight:115.13 g/mol
Merck Index:13,07871
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:205-702-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:1500 g/l (20 °C)
- Melting Point:220 - 222 °C (decomposition)
- Molar Mass:115.13 g/mol
- Bulk Density:500 kg/m3
- Density:1.35 - 1.38 g/cm3 (25 °C)
- pH:5 - 7 (100 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together