ddATP [2',3'-Dideoxyadenosine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Catalog # AATB17209
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:1 µmol
- Application:
- Description:ddATP [2',3'-Dideoxyadenosine-5'-triphosphate] *10 mm in ddH₂O*
- Environmentally Preferable:
Specifications
About this item
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
- In-house quality control
- Useful for sanger sequencing applications
Caution: For research use only.