D-Panthenol ≥98% USP
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: Dexpanthenol, (R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
CAS Number:81-13-0
:
Formula:C₉H₁₉NO₄
MDL Number:MFCD00065006
Molecular Weight:205.25 g/mol
Storage Temperature:Ambient
EINECS:201-327-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:98% Min.
Specification Test Results
Frequently Bought Together