Order Entry
Netherlands
ContactUsLinkComponent
Cleanroom lab coats
undefined
Cleanroom lab coats
  • Catalog No:
  • ALSO1037468-L
  • ALSO1037470-3XL
  • ALSO1037468-XXL
  • ALSO1037468-XL
  • ALSO1037470-XS
  • ALSO1037468-3XL
  • ALSO1037470-M
  • ALSO1037470-S
  • ALSO1037468-XS
  • ALSO1037470-XL
  • ALSO1037470-L
  • ALSO1037468-M
  • ALSO1037468-S
  • ALSO1037470-XXL
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Risk area:
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Fabric:
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Alsistrong® (98% polyester, 2% carbon)
  • Weight:
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
  • 118 g/m²
Frequently Bought Together