PCR tube strips and dome cap strips, Axygen®
Supplier: Corning
Certificates
About this item
These polypropylene tube strips and cap strips are specifically designed for PCR use and are ideal for thermal transfer applications.
- Made from highly polished molds
- Free of lubricants, dyes, heavy metals, and fillers
The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling. Tubes offer a maximum RCF of 4000 g. Tubes and caps are certified by lot number to be RNase-, DNase-, and pyrogen-free.
Ordering information: Tube strips and cap strips are ordered separately. For Real Time PCR applications, select the clear tube strips and the Real Time PCR cap strips.
Specifications
- Catalog No:
- AXYGPCR-02CP-C-100
- AXYGPCR-0208-C-100
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa